Learn More
Benchmark Scientific™ UV-C Indicator Card for Research Use Only
Click to view available options
:
Benchmark Scientific™ UV-C Indicator Card
Description
- Changes color in the presence of UV-C
- Used to verify operation of UV-C lamps
- Double sided
- For research use only
Specifications
Specifications
| For Use With (Application) | Confirm Operation of UV-C Lights |
| For Use With (Equipment) | UV-Clave |
| Model | Double-Sided Card |
| Sterilization Indicator | Color Change Indicator |
| Sterility | Non-sterile |