Learn More
IKA™ UTL 25 Digital Inline ULTRA-TURRAX™ with UK Plug
Product Code. 17593596 Shop All IKA Products
Click to view available options
Speed:
3000 to 25,000 rpm
Unit Size:
Each
Product Code. 17593596
Brand: IKA™ 0010006858
Only null left
Add to basket
Add to basket
UTL 25 digital Inline ULTRA-TURRAX™ for circulation or flow-through processing in the laboratory.
  • Simple, compact and sturdy modular design
  • Sterilizable, autoclave-compatible
  • Table-top or stand-supported device, low space requirement
  • Easy disassembly
  • Large delivery capacity of 4.4 to 11.6 L/min. with open outlet (the mounting of a valve can reduce the flow rate)
  • For air-free, sterile, and inline suspension, emulsifying and deagglomeration
  • For vacuum or pressurized operation (up to 6 bar)
  • If the DK 25.11 is used, air induction is also prevented in batch operation
  • Not self-priming
  • A pump can be integrated between intake nozzle and vessel. As a result, viscous fluids can be processed
  • Not suitable for continuous operation or cyclical continuous operation
  • The hoses used should have an inner diameter size of 13 mm (inlet) and 9 mm (outlet) respectively to fit the DK 25.11 flow chamber
TRUSTED_SUSTAINABILITY
Specifications
Speed 3000 to 25,000 rpm
Voltage 230 V
Hertz 50/60 Hz
Capacity (Metric) 26 mL
Diameter (Metric) Shaft 13 mm
Dimensions (D x W x H) 450 x 120 x 100 mm
Length (Metric) Probe 175 mm
For Use With (Application) Circulation or Flow-Through Processing
Noise Level 73 dB
Plug Type UK
Weight (Metric) 3.8 kg