Learn More
Cole-Parmer™ Economical Utility Gauge
Product Code. 11739829 Shop All Cole-Parmer Products
Change view
Click to view available options
Pressure Range:
0 to 100 psi
0 to 15 psi
0 to 160 psi
0 to 30 psi
0 to 300 psi
0 to 60 psi
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pressure Range Unit Size
11739829 0 to 15 psi Each
11759829 0 to 60 psi Each
11749829 0 to 30 psi Each
11769829 0 to 100 psi Each
11779829 0 to 160 psi Each
11799829 0 to 300 psi Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11739829 Supplier Cole-Parmer™ Supplier No. 6835518
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
The Economical Utility Gauge is a high-quality, low cost gauge designed for industrial processes.
  • Utility gauges are designed to replace more expensive digital or commercial gauges
Specifications
Certifications/Compliance CE Compliance
For Use With (Application) High-quality, low cost gauges designed for industrial processes
Size (English) Fitting 0.1 in. NPT(M), Lower
Size (Metric) Fitting 3.17 mm NPT(M), Lower
Accuracy 3-2-3% Full-scale
Display Type Analog
Pressure Range 0 to 15 psi
Product Type Utility Pressure Gauge