Learn More
3M™ Doodlebug™ Utility Pad Holder
Product Code. 17198640
Click to view available options
Color:
Brown
Unit Size:
Pack of 10
Product Code. 17198640
Brand: 3M™ Doodlebug™ 7000086421
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A durable floor pad holder with swivel head for use with the 3M™ Doodlebug™ Floor Edge Cleaning Kits. Pad holder is typically used with a handle for cleaning without bending over. Cleaning pads and broom handle are sold separately.
  • Floor pad holder with swivel head for reaching hard to get at places
  • Typically used with a handle for cleaning without bending over
  • Cleaning pads & broom handle sold separately
TRUSTED_SUSTAINABILITY
Specifications
Product Type Utility Pad Holder
Color Brown
Dimensions (L x W) 254 x 118 mm
For Use With (Equipment) 3 M™ Doodlebug™ Floor Edge Cleaning Kits
Length (English) 9 in.
Length (Metric) 254 mm
Quantity 10 Pk.
Width (English) 3.75 in.
Width (Metric) 118 mm