Learn More
Olympus™ Ushio™ USH-103D Long Life Short Arc Mercury Lamp
Product Code. 10321470
Change view
Click to view available options
USH-103D Long Life Short
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10321470 USH-103D Long Life Short
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10321470 Supplier Olympus™ Supplier No. N1217800
  • ONLINE EXCLUSIVE PRICE
    1015.00€
    412.55€ / Pack of 1
    Save 602.45€
    59% Off
 Restricted Item
Estimated Shipment: 15-10-2026
With an enhanced electrode design for precise positioning and high arc stability.
  • Improved manufacturing process enables extended life (300 hours)
  • Ushio-developed product with tightly controlled ISO quality processes
  • Designed for use in all 100W fluorescent microscope systems
  • Characterized by tight plasma discharge, high luminance and ultra-stable arc
  • 100 watts, 20 volts
Specifications
Type Mercury Short-arc lLamp
Description 100 W, 20 V