Learn More
Memmert™ USB User-ID Stick (with User-ID License) for IN450plus
Product Code. 17567755 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN450plus Incubator
Unit Size:
Each
Product Code. 17567755
Brand: Memmert™ IN450plusB33170
Only null left
Add to basket
Add to basket
USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
  • Oven-linked authorization License (User-ID-program) on Memory-stick
  • Prevents undesired manipulation by unauthorised third parties
  • When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
For Use With (Equipment) IN450plus Incubator
Type USB Flash Drive
Includes User ID Licence