Learn More
Memmert™ USB User-ID Stick (with User-ID License) for IF30plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF30plus Incubator
Unit Size:
Each
Description
- Oven-linked authorization License (User-ID-program) on Memory-stick
- Prevents undesired manipulation by unauthorised third parties
- When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Specifications
| For Use With (Equipment) | IF30plus Incubator |
| Type | USB Flash Drive |
| Includes | User ID Licence |