Learn More
Memmert™ Usb User-Id Stick (With User-Id Licence), For Vo49
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Units With TwinDISPLAY
Unit Size:
Each
Description
Includes
User-ID License
- USB User-ID stick (with User-ID licence)
- Oven-linked authorisation licence (User-ID-programme) on Memory-stick
- Prevents undesired manipulation by unauthorised third parties
- When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
Specifications
| Description | Only for units with TwinDISPLAY |
| For Use With (Equipment) | Units With TwinDISPLAY |
| Type | USB Stick |