missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ursolic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Ursolic Acid |
| CAS | 77-52-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C30 H48 O3 |
| Quantity | 100 mg |
| Synonym | Urs-12-en-28-oic acid','3-hydroxy-','(3β)- (9CI','ACI), (3β)-3-Hydroxyurs-12-en-28-oic acid (ACI), Urs-12-en-28-oic acid','3β-hydroxy- (8CI), (+)-Ursolic acid, 3β-Hydroxyurs-12-en-28-oic acid, Bungeolic acid, Malol, Merotaine, Neoage UR, NSC 167406, NSC 4060, Prunol, Ursolic acid, Ursolisome, Urson, β-Ursolic acid |
| InChI Formula | InChI=1S/C30H48O3/c1-18-10-15-30(25(32)33)17-16-28(6)20(24(30)19(18)2)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h8,18-19,21-24,31H,9-17H2,1-7H3,(H,32,33)/t18-,19+,21+,22-,23+,24+,27+,28-,29-,30+/m1/s1 |
| SMILES | C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(=O)O |
| IUPAC Name | (1S,2R,4aS,6aR,6aS,6bR,8aR,10S,12aR,14bS)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid |
| Molecular Weight (g/mol) | 456.7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction