missing translation for 'onlineSavingsMsg'
Learn More

Ursodeoxycholic Acid, TRC

Product Code. 17903185
Change view
Click to view available options
Quantity:
5 mg
1 g
5 g
Unit Size:
1g
5g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903185 1 g 1g
17981415 5 g 5g
17963625 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17903185 Supplier Toronto Research Chemicals Supplier No. TRCU8500001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-13-2
Molecular Formula C24 H40 O4
Molecular Weight (g/mol) 392.57
Synonym Ursodeoxycholic acid, 3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid, 3α,7β-dihydroxy- (8CI), 5β-Cholanic acid, 3α,7β-dihydroxy- (7CI), 17β-(1-Methyl-3-carboxypropyl)etiocholane-3α,7β-diol, 3α,7β-Dihydroxy-5β-cholan-24-oate, 3α,7β-Dihydroxy-5β-cholan-24-oic acid, 3α,7β-Dihydroxy-5β-cholanic acid, 3α,7β-Dihydroxy-5β-cholanoic acid, 3α,7β-Dihydroxycholanic acid, 7β-Hydroxylithocholic acid, Actigall, Adursal, Arsacol, Cholit-Ursan, Delursan, Desocol, Desol, Destolit, Deursil, Litursol, Lyeton, NSC 683769, Paptarom, Solutrat, UDCA, Urdes, Ursacol, Ursic acid, Urso, Ursobilin, Ursochol, Ursocholic acid, deoxy-, Ursocol, Ursodamor, Ursodeoxycholic acid, Ursodesoxycholic acid, Ursodiol, Ursofalk, Ursolvan, Ursosan
IUPAC Name (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1
Formula Weight 392.2927
Percent Purity >95%
Chemical Name or Material Ursodeoxycholic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.