Learn More
Gilac™ Urine Jar
Product Code. 17988971
Change view
Click to view available options
Volume (Metric):
2 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17988971 2 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17988971 Supplier Gilac™ Supplier No. G122421
  • ONLINE EXCLUSIVE PRICE
    9.45€
    6.20€ / Pack of 1
    Save 3.25€
    34% Off
Estimated Shipment: 09-12-2026
Urine Jar is graduated, very stable even when empty, and benefits from a screw-on lid. Easy to clean.
  • 2 L urine Jar.
  • Outstanding durability even in the intensive use cycle.
  • Screw lid, bocal with graduation.
  • Researched product ergonomics ensure optimum cleaning.
  • Suitable for hospital use.
  • Product can be disinfected in a bedpan washer.
Specifications
Product Type Urinal
Color Transparent
Material Polyethylene
Sterility Non-sterile
Disposable Reusable
Graduated Yes
Volume (English) 0.5 gal.
Volume (Metric) 2 L