Learn More
Tocris Bioscience™ uPSEM 817 tartrate
GREENER_CHOICE
Product Code. 18005742
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Product Code. 18005742
Brand: Tocris Bioscience™ 6866/5
Highly potent and selective PSAM4-GlyR and PSAM4-5-HT3 chimeric ion channel agonist; brain-penetrant
uPSEM 817 tartrate is a selective ultrapotent PSEM (uPSEM) agonist for α7L131G,Q139L,Y217F-GlyR (PSAM4-GlyR) and PSAM4-5-HT3 chimeric ion channels (EC50 values are 0.3 and 0.5 nM, respectively). Suppresses firing of layer 2/3 cortical neurons expressing PSAM4-GlyR in brain slices. Increases contralateral rotation in mice expressing PSAM4-GlyR unilaterally in the substantia nigra reticulata (LED 0.1 mg/kg).
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Propoxy-7,8,9,10-tetrahydro-6 H-6,10-methanoazepino[4,5-g]quinoxaline L-tartrate
CAS 2341833-14-3
Quantity 5 mg
Target Pharmacologically Selective Effector Molecules/PSEM
Molecular Formula C16H19N3O.C4H6O6
Formula Weight 419.43
Purity 98%