Learn More
Berthold Technologies Upper Part For DeWit Tubes
Description
- The turntable for NightSHADE enables sequential and scheduled acquisition of up to 18 DeWitt tubes (in groups of three)
- The component represents the upper part of the turntable.
- Used together with the turntable base
Specifications
Specifications
| Type | Specimen Tray |
| For Use With (Application) | Imaging of Seedlings |
| For Use With (Equipment) | Imaging System |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?