Learn More
Idex Tubing Sleeves
Ensure good port connection with capillary tubing
Brand: Idex F-181
Additional Details : Weight : 0.00045kg
Description
Ensure good port connection with capillary tubing
- Maintain concentric connection with minimal dead volume
- Color coded for easy identification
Sleeves are not permanently attached to a ferrule so they can easily adapt to varying tubing pocket depths minimizing dead volume. Sleeves are designed to be used with 1/16 in. OD, 10-32 threaded fittings to adapt capillary tubing to standard coned ports. Sleeves require a wrench tightened nut to achieve proper sealing and withstand temperatures up to 257°F (125°C) and pressures up to 6000 psi (414 bar).
Specifications
Sleeve | |
0.006 to 0.010 in. | |
125-165 μm O.D. Tubing |
Yellow | |
180 μm | |
PEEK |