Learn More
Macherey-Nagel™ Untreated Capillary
Product Code. p-8012446
Click to view available options
Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Pack of 10
Pack of 3
Product Code. 10266271
Brand: Macherey-Nagel™ 723101.25
Only null left
Specifications
Diameter (Metric) 0.4 mm
Type Untreated Capillary
Diameter (Metric) Inner 0.25 mm
For Use With (Application) Preparation of Capillary Columns Analysis, Preparation of Capillary Columns, LC
Length (Metric) 25 m
Quantity 1 Each