Learn More
Macherey-Nagel™ Untreated Capillary
Click to view available options
Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Pack of 10
Pack of 3
Specifications
Specifications
| Diameter (Metric) | 0.5 mm |
| Type | Untreated Capillary |
| Diameter (Metric) Inner | 0.32 mm |
| For Use With (Application) | LC Analysis, Preparation of Capillary Columns, LC |
| Length (Metric) | 25 m |
| Quantity | 1 Each |