Learn More
Macherey-Nagel™ Untreated Capillary
Product Code. 10388491
Click to view available options
Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Pack of 10
Pack of 3
Product Code. 10388491
Brand: Macherey-Nagel™ 723792.1
Specifications
Diameter (Metric) 0.4 mm
Type Untreated Capillary
Diameter (Metric) Inner 0.1 mm
For Use With (Application) LC Analysis, LC
Length (Metric) 1 m
Quantity 1 Each