Learn More
Macherey-Nagel™ Unmodified Cellulose MN 100
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Specifications
Specifications
| Grade | Standard |
| Quantity | 5 kg |
| Surface Area (Metric) | ∼6500 cm2/g |
| Type | Unmodified Cellulose MN 100 |