Learn More
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
12 mm Shaft
16 mm Shaft
6 mm Shaft
8 mm Shaft
Unit Size:
Pack of 5
Specifications
Specifications
| Description | Fisherbrand Universal Stirrer Guide Replacement Lock Nut Pack |
| For Use With (Equipment) | 12 mm Shaft |
| Includes | Blue Lock Nut, Ferrule & Shaft Seal |
| Quantity | 1 |
| Temperature (Metric) | -200°C to +280°C |