Learn More
asecos™ Universal Sorbents Sock
Product Code. 17896416 Shop All Asecos Products
Click to view available options
Length (Metric):
1.2 m
3 m
Unit Size:
Pack of 15
Pack of 6
Product Code. 17896416
Brand: asecos™ 11347
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Universal Sorbents Sock is ideal for the safe absorption of oils, water, coolants and non-aggressive chemicals.
  • Very strong durability
  • Suitable for heavy and long-lasting use
  • Tightly woven fibers, high abrasion resistance and low-lint surface
  • Outstanding absorption performance
  • Large absorption capacity (up to 16 time of own weight)
  • Light for smaller leakages and prevention
  • Heavy for larger leakages or absorption of puddles
Specifications
Color White
Material Heavy-duty Cloth
Type Universal Sorbent Sock
Length (English) 9.84 ft.
Length (Metric) 3 m
Width (English) 2.95 in.
Width (Metric) 75.00 mm
Weight (English) 13.23 lb.
Weight (Metric) 6 kg