Learn More
Fisherbrand™ Universal pH Indicator Paper
Product Code. 11557392 Shop All Fisherbrand Products
Click to view available options
pH Range:
0.5 to 5
0.5 to 5.5
1 to 11
1 to 14
4 to 7
5 to 9
6.5 to 10
9 to 13
Format:
Roll
Tube
Length (Metric):
5 m
50 mm
Quantity:
1 Roll
10 Pack
4 Rolls
Unit Size:
Each
Pack of 4
Product Code. 11557392
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Gives rough knowledge of pH.
  • Provides rapid and precise readings
  • Includes color scale printed on cover for convenience
Applications:
  • Used for general analysis in chemical, school laboratories
Specifications
pH Range 9 to 13
Format Roll
Quantity 4 Rolls
Type Universal pH indicator Paper
Length (Metric) 5 m
Width (Metric) 7 mm