Learn More
Fisherbrand™ Universal pH Indicator Paper
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
Brand: Fisherbrand™
Additional Details : Weight : 0.06120kg
Description
- Filter paper soaked with one or several colored indicators of pH
- Economic range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows color changes in steps of pH2
Specifications
6.4 to 8 | |
4 Rolls | |
5 m |
Roll | |
Grade 911 | |
7 mm |