Learn More
Cole-Parmer™ Union Threaded Elbow
Threaded fittings offer more permanent installation
Brand: Cole-Parmer™ 06546-36
Additional Details : Weight : 0.80000kg
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Union Threaded Elbow | |
Not Autoclavable | |
3/4 in. | |
3/4 in. NPTF x 3/4 in. NPTF |
NPT(F) Connection | |
PFA | |
Ethylene Oxide | |
Translucent |