Learn More
Cole-Parmer™ Union Threaded Elbow
Threaded fittings offer more permanent installation
78.60€ - 168.00€
Specifications
Product Type | Union Threaded Elbow |
---|---|
Description | NPT(F) Connection |
Autoclavable | Not Autoclavable |
Material | PFA |
Sterilization Method | Ethylene Oxide |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
11848033
|
Cole-Parmer™
06546-32 |
1/8 in. NPTF x 1/8 in. NPTF |
78.60€
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
11868033
|
Cole-Parmer™
06546-34 |
3/8 in. NPTF x 3/8 in. NPTF |
121.00€
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
11878033
|
Cole-Parmer™
06546-35 |
1/2 in. NPTF x 1/2 in. NPTF |
143.00€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
11888033
|
Cole-Parmer™
06546-36 |
3/4 in. NPTF x 3/4 in. NPTF |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Union Threaded Elbow | |
Not Autoclavable | |
Ethylene Oxide |
NPT(F) Connection | |
PFA | |
Translucent |