Learn More
Hirschmann™ Union Nut
Product Code. 10004732
Change view
Click to view available options
Color:
Red
Turquoise
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10004732 Turquoise Each
10644893 - Each
10710654 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10004732 Supplier Hirschmann™ Supplier No. 9903302
  • ONLINE EXCLUSIVE PRICE
    20.63€
    13.50€ / Pack of 1
    Save 7.13€
    34% Off
Estimated Shipment: 29-10-2026
Retainer rings for use with the Pipetus™ pipette filler line.
  • Available in a range of colors
  • Aids in liquid handling
  • Accessory item
Specifications
Color Turquoise
Type Union Nut
For Use With (Equipment) Pipetus