Learn More
Abeba™ UNI6 1781 Shoes
Product Code. 15697737
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15697737 35 1 pair
15607747 36 1 pair
15617747 37 1 pair
15627747 38 1 pair
15637747 39 1 pair
15647747 40 1 pair
15657747 41 1 pair
15667747 42 1 pair
15677747 43 1 pair
15687747 44 1 pair
15697747 45 1 pair
15607757 46 1 pair
15617757 47 1 pair
15627757 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15697737 Supplier Abeba™ Supplier No. 1781/35
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Steel toe-cap provides extra protection. Special lining with silver ions absorbs resperation allowing the foot to breathe.
  • CE EN ISO 20345:2011 S2 SRC
  • A-micro
  • Black
  • Steel toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350122)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver Point
  • Instep area with lacing
  • **Size 35 with replaceable acc Wave insole (art. 3576)
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color Black
Material A-micro
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1781
Toe Material Steel