Learn More
Duran™ Ungraduated Measuring Cylinders
Product Code. 15513162 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15513162 250 mL Pack of 10
15503162 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15513162 Supplier Duran™ Supplier No. 213913679
  • ONLINE EXCLUSIVE PRICE
    151.20€
    131.64€ / Pack of 10
    Save 19.56€
    12% Off
Estimated Shipment: 30-10-2026
These measuring cylinders are for holding and measuring varying amounts of liquids in the laboratory.
  • Without graduation
  • Volume content tolerances for measuring and mixing cylinders conform to DIN and ISO accuracy limits
  • Robust, virtually inert and exceptionally resistant to high temperatures
  • Outstanding corrosion resistance
  • Manufactured from borosilicate glass 3.3
  • Large hexagonal base has three knobs to increase stability and prevents cylinder from rolling
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Certifications/Compliance DIN EN ISO 4788
For Use With (Application) Laboratories
Base Type Hexagonal
Quantity 10/Pk.
Product Type Ungraduated Measuring Cylinder