Learn More
Haldenwanger™ Glazed Porcelain Mortar without Pestle
Product Code. 12313328
Change view
Click to view available options
Capacity (Metric):
70 mL
75 mL
100 mL
170 mL
250 mL
400 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12313328 75 mL Each
12323328 100 mL Each
10053504 170 mL Each
12343328 250 mL Each
12353328 400 mL Each
12303328 70 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12313328 Supplier Haldenwanger™ Supplier No. L 55/1/RAUH
Only null left
Add to basket
Add to basket
Use these mortars for a variety of laboratory applications. Haldenwanger™ Unglazed Porcelain Mortars are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications
Diameter (Metric) Inner 70mm
Product Type Mortar
Material Porcelain
Height (Metric) 45mm
Finish Unglazed
Capacity (Metric) 75 mL