Learn More
BICASA Undercounter Drawer Cabinet on Casters
Product Code. 30057610 Shop All BICASA Products
Change view
Click to view available options
Color:
Anthracite
Blue
Light Gray
Red
Dimensions (D x W x H):
600 x (500 + 20) x 630 mm
600 x (500 + 20) x 760 mm
900 x (500 + 20) x 760 mm
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (D x W x H) Unit Size
30057610 Red 600 x (500 + 20) x 630 mm Each
30057571 Light Gray 600 x (500 + 20) x 630 mm Each
30057583 Anthracite 600 x (500 + 20) x 630 mm Each
30057581 Blue 600 x (500 + 20) x 630 mm Each
30057561 Light Gray 600 x (500 + 20) x 760 mm Each
30057562 Anthracite 600 x (500 + 20) x 760 mm Each
30057566 Blue 600 x (500 + 20) x 760 mm Each
30057589 Red 600 x (500 + 20) x 760 mm Each
30057601 Light Gray 900 x (500 + 20) x 760 mm Each
30057599 Anthracite 900 x (500 + 20) x 760 mm Each
30057579 Blue 900 x (500 + 20) x 760 mm Each
30057563 Red 900 x (500 + 20) x 760 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 30057610 Supplier BICASA Supplier No. TN.DW.0600.630R
  • ONLINE EXCLUSIVE PRICE
    572.00€
    566.00€ / Pack of 1
    Save 6.00€
    1% Off
View all promotions
Estimated Shipment: 26-10-2026
Undercounter Drawer Cabinet on Casters with fire-retardant particleboard body and drawers with soft closing and single lock.
  • Undercounter cabinets on casters
  • Metal frame base with 4 swivel casters (Polyamide case and thermoplastic rubber treads), 2 with lockable with brakes - Height mm 100
  • Central locking system as standard
  • Anti-tilt safety system to open only one drawer at a time
  • Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
Specifications
Product Type Mobile Storage Cabinet
Color Red
Cabinet Type Underbench cabinet
Dimensions (D x W x H) 600 x (500 + 20) x 630 mm
No. of Doors 1
No. of Drawers 3
Width (English) 23.6 in.
Width (Metric) 600 mm
With Casters Yes
Material Melamine