missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecanoic-d21 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Undecanoic-D21 Acid |
| CAS | 60658-40-4 |
| Molecular Formula | C11HD21O2 |
| Quantity | 0.5 g |
| Synonym | Undecanoic-d21 acid (9 CI), Undecanoic acid D21, Undecanoic acid-d21, Perdeuteroundecanoic acid |
| InChI Formula | InChI=1 S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(O)=O |
| IUPAC Name | undecanoic-d21 acid |
| Molecular Weight (g/mol) | 207.42 |
| Formula Weight | 207.29 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction