missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UNC 2881

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
UNC 2881 is a potent Mer kinase inhibitor. Displays >55-fold selectivity for Mer kinase over other TAM receptor tyrosine kinase family members (IC50 values are 4.3 nM, 0.25 μM and 0.36 μM for Mer, Tyro3 and Alk, respectively). Inhibits Mer phosphorylation in vitro. Also inhibits platelet aggregation in response to Type I collagen.
Specifications
Specifications
| Chemical Name or Material | 2-(Butylamino)-4-[(trans-4-hydroxycyclohexyl)amino]-N-[[4-(1H-imidazol-1-yl)phenyl]methyl]-5-pyrimidinecarboxamide |
| CAS | 1493764-08-1 |
| Quantity | 5 mg |
| Molecular Formula | C25H33N7O2 |
| Formula Weight | 463.58 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido