missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UNC 0638

Product Code. 18004072
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18004072 10 mg 10mg
18025252 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 18004072

Brand: Tocris Bioscience™ 4343/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective G9a and GLP inhibitor

Selective inhibitor of G9a and GLP histone lysine methyltransferases (IC50 values are < 15 nM and 19 nM for G9a and GLP respectively, and > 10000 nM for a range of other histone methyltransferases). Potently inhibits dimethylation of H3K9 in MCF-7 cells (IC50 = 70 nM). Cell permeable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1255517-77-1
Molecular Formula C30H49N5O3

Specifications

Quantity 10 mg
Formula Weight 509.73
Purity >99%
Molecular Weight (g/mol) 527.754
Chemical Name or Material UNC 0638
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.