Learn More
Bandelin Electronic™ Tickopur™ R 33 Ultrasonic Cleaning Fluid
Product Code. 11823482
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11823482 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11823482 Supplier Bandelin Electronic™ Supplier No. 831
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A universal cleaner for the laboratory, service and industry. Bandelin Electronic™ Tickopur™ R 33 Ultrasonic Cleaning Fluid provides excellent protection against corrosion.
  • Concentrated
  • Mildly alkaline, pH 9.9 (1%)
  • Dosage 1-5%
  • Cleans and disinfects items such as technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
pH 9.9
Concentration 1%
Container Type Jerrican
Volume (Metric) 5 L