Learn More
Diversey Ultraclean VK3, 20L, General purpose liquid detergent for manual use
Brand:  Diversey 100852224
Ultraclean VK3, 20L, General purpose liquid detergent for manual use
Product Code. 17653225
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Ultraclean VK3
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Propan-2-ol, alkyl alcohol ethoxylate, sodium cumenesulphonate, sodium hydroxide