Learn More
IKA™ ULTRA-TURRAX™ Tube Drive P Control Disperser
Product Code. 16298661 Shop All IKA Products
Change view
Click to view available options
Speed:
400 to 8000 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Unit Size
16298661 400 to 8000 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16298661 Supplier IKA™ Supplier No. 0025005981
  • ONLINE EXCLUSIVE PRICE
    1760.00€
    1669.61€ / Pack of 1
    Save 90.39€
    5% Off
In Stock
Assures the best stirring, dispersing, grinding and homogenisation results and has been devised in particular for the safe processing of infectious, toxic or odour-intensive samples.
  • Disposable tubes
    The sealable disposable tubes have been optimized for your application.
    They reduce the amount of cleaning required and rule out any risk of cross-contamination.
  • Programmable test conditions
    Time, energy, volume: You specify and systematically program the test conditions and store frequently used parameters in your library.
  • Adjustable change in direction of rotation
    When, and how often, the direction of rotation should be changed is very easily set using the clear and multilingual menu.
  • Turbo button
    Even more intensive mixing, dispersing or grinding at your fingertips: All of this is achieved with the turbo button. Over short periods, rotational speeds of up to 8,000 rpm can be achieved.
TRUSTED_SUSTAINABILITY
Specifications
Speed 400 to 8000 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 36 W
Wattage 36 W
Dimensions (L x W x H) 178 x 122 x 54 mm
Material Stainless Steel
Display LED
For Use With (Equipment) Multi Drive
Timer Yes
Viscosity 5000 mPas
Weight (Metric) 1.3 kg