Learn More
Thermo Scientific™ Ultra Pure Water Dispensing and Shut-Off Taps
Use these PVC taps with high-purity water. Available with straight or angled nozzle, for use with SDI systems (behind the tanks).
323.00€
Product Code | Brand | Description | Inlet Connection NPTF (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Inlet Connection NPTF (English) | Price | Quantity & Availability | ||||
15453452
|
Thermo Scientific™
50155456 |
Pure water outlet, straight | R ¾ in. union nut |
323.00€
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
Description
Use these PVC taps with high-purity water. Available with straight or angled nozzle, Thermo Scientific™ Ultra Pure Water Dispensing and Shut-Off Taps are used with SDI systems, behind the tanks.
- For use with high purity water
- Available with straight or angled (90°) nozzle
- Material: PVC
Specifications
PVC |