Learn More
3M™ Ultra Durable Floor Marking Tape 971
Product Code. 17267800
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Unit Size:
1 roll
Product Code. 17267800
Brand: 3M™ 7100167106
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ultra Durable Floor Marking Tape 971
  • Excellent abrasion resistance
  • Withstands lifting from forklift traffic
  • Bright colors for high visibility
  • Resists moisture and many chemicals
  • Sticks on contact and stays where you put it
  • Promotes clean removal without solvents and scraping
  • Available in a variety of colours
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 51 mm x 33 m
Length (Metric) 32.91 m
Width (Metric) 50.8 mm
Color Green