missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP-2/5/6 Mouse anti-Human, Alexa Fluor 488, Clone: 165903, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1298G-100UG
This item is not returnable.
View return policy
Description
ULBP-2/5/6 Monoclonal specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry.Specifications
| ULBP-2/5/6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BZM5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ULBP-2 and human RAET1L/ULBP-6 in direct ELISA and stains cells transfected with human ULBP-2, human ULBP-5 or human RAET1L/ULBP-6 in flow cytometry. In direct ELISA, 11% cross-reactivity with recombinant human ULBP5 is observed. It does not stain cells transfected with human ULBP-1 or human ULBP-3 in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 165903 | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ALCAN-alpha, N2DL2, NKG2DL2, RAET1G, RAET1H, RAET1L, UL16 binding protein 2, ULBP2, ULBP5, ULBP6 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-2 Accession # Q9BZM5 | |
| RUO | |
| 80328 | |
| Human | |
| IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)