missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UBP 310
GREENER_CHOICE

Product Code. 18086002
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18086002

Brand: Tocris Bioscience™ 3621/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GluK1-selective kainate antagonist

UBP 310 is a GluK1 (formerly GLUK5) kainate receptor antagonist (IC50 = 130 nM); also blocks recombinant homomeric GluK3 (forrmerly GLUK7) receptors. Displays 12,700-fold selectivity for GluK1 (formerly GLUK5) over GluK2 (formerly GLUK6). Exhibits no activity at mGlu group I or NMDA receptors at concentrations of up to 10 μM. Apparent KD value is 18 ± 4 nM for depression of kainate responses on the dorsal root.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type UBP 310
Purity >98%
Molecular Weight (g/mol) 353.35
Inhibitors UBP 310

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.