missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ UBL5 Recombinant Protein

Brand:  Abnova™ H00059286-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133153

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH07053
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
34
8
Glutathione Sepharose 4 Fast Flow
25 ug
MIEVVCNDRLGKKVRVKCNTDDTIGDLKKLIAAQTGTRWNKIVLKKWYTIFKDHVSLGDYEIHDGMNLELYYQ
FLJ46917/HUB1/MGC131795
UBL5
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
59286
UBL5 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
UBL5
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers