missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ubiquitin Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| For Use With (Application) | ELISA |
| Formulation | Prepared from bovine erythrocytes. |
| Gene ID (Entrez) | 6233 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 8.5 kDa |
| Quantity | 25 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Gene Alias | RPS27A, UBA52, UBB ubiquitin B, UBC |
| Gene Symbol | RPS27A |
| Reconstitution | Reconstitute with 25 mL distilled water. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution. For long term storage the addition of 0.09% sodium azide is recommende) |
| Research Category | Alzheimers Research, Autophagy, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction