Learn More
SI Analytics™ Ubbelohde Viscometers for Manual Measurements
Product Code. p-7160705 Shop All SI Analytics Products
Click to view available options
Viscosity Range:
0.8 to 5 cSt
50 to 500 cSt
Approximate Constant:
0.005
0.5
Unit Size:
Each
Product Code. 12008163
Brand: SI Analytics™ 285400029
SI Analytics™ Ubbelohde Viscometers for Manual Measurements feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high-temperature or low temperature measurements
  • Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • All viscometers are provided with ring marks
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • The recommended minimum flowthrough time is 200 seconds
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ubbelohde Viscometer
Viscosity Range 0.8 to 5 cSt
Calibrated Yes
Description 0.53 mm diameter
Approximate Constant 0.005
Certifications/Compliance DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Material Glass
Length (Metric) 290 mm
For Use With (Application) Manual measurements
Volume (Metric) 15 to 20 mL