Learn More
uvex U-Chem 3100 Chemical Protection Gloves
Product Code. p-7230507 Shop All Uvex Products
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
Product Code. 17240521
Brand: uvex 6096808
Only null left
Add to basket
Add to basket
Gloves are constructed of nitrile butadiene rubber (NBR) with a cotton interlock liner, an additional coating for enhances grip capabilities when handling wet or oily objects.
  • Nitrile butadiene rubber (NBR) chemical protection gloves
  • An additional coating enhances grip capabilities when handling wet/oily objects
  • Ideal for a wide variety of applications that require mechanical protection
TRUSTED_SUSTAINABILITY
Specifications
Applications Laboratory
Coating Material NBR
Color Black
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 8
EN388 Standard 4121X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMO
For Use With (Application) Automotive, Chemical Handling, Metal Work, Mechanics, and Sand Blasting
Gender Unisex
Glove Cuff Style Gauntlet
Series U-Chem 3100