Learn More
uvex U-Chem 3100 Chemical Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
8
9
10
11
Unit Size:
10 pairs
Description
- Nitrile butadiene rubber (NBR) chemical protection gloves
- An additional coating enhances grip capabilities when handling wet/oily objects
- Ideal for a wide variety of applications that require mechanical protection
Specifications
Specifications
| Applications | Laboratory |
| Coating Material | NBR |
| Color | Black |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Chemical-resistant Gloves |
| Size | 9 |
| EN388 Standard | 4121X |
| EN ISO 374-1 | Type A |
| EN ISO 374-1 CHEMICAL LETTER CODES | AJKLMO |
| Show More |