missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ U-2 OS Nuclear Cell Lysate

Product Code. 18582980 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18582980 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18582980 Supplier Novus Biologicals™ Supplier No. L022V4
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nuclear Extract Lysate

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Use it directly for immuno-precipitation, or heat lysate with SDS gel loading buffer to 95C for 5 minutes followed by rapid cooling for western blot application. If dissociating conditions are required, add reducing agent prior to heating.
Quantity 0.05 mg
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Target U-2 OS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.