missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F Plus with Attached Gloves Model CHZ5
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 10
Description
- DuPont™ Tychem™ 6000 F Plus with attached non-dissipative undergloves, TFCHZ5TGY08. Hooded coverall. Stitched and over-taped seams. Double cuffs with attached chemical barrier gloves on inner cuff. Elastication at wrists, ankles, face, and waist. Double zippers and double flaps. Outer flap with adhesive tape. Grey.
- Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek ™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
- Tychem™ 6000 F garments are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | HDPE |
| Color | Gray |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction