Description
- Elasticated top and fixation ties
- Provides an ideal balance of protection, durability and comfort
- Silicon non-added
- Accessory
Specifications
Specifications
| Size | One Size |
| Material | Polyethylene |
| Color | White |
| Sole Type | Slip-resistant |
| Model | POBA Overboot |
| Seam Type | Stitched |
| For Use With (Application) | Pharmaceutical Handling, Chemical Protection, Lead and Asbestos Abatement/Remediation, General Maintenance/Operations, Spray Painting and General Clean-up, Amongst Many Others. |
| Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126, EN 1149-1 |