missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 400 Dual TG Model TD148S
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Description
- DuPont™ Tyvek™ 400 Dual TG. Hooded coverall combining Tyvek™ at the front with a SMS nonwoven on the back panel. Available in sizes SM to 3X. Attached Tyvek™ under gloves. Attached Tyvek™ socks. 2-piece hood and gusset for optimal fit. Elasticated hood and waist. White.
- Tyvek™ front provides an ideal balance of protection, durability and comfort. It is permeable to both air and water vapor, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size on the Tyvek™ front. It is ultra-low-linting and antistatically treated. Silicone non-added. The large, breathable back panel, made of SMS nonwoven, offers lower protection against particles and light water-based splashes, yet high levels of comfort.
- Tyvek™ 400 Dual TG is designed for specific applications that demand comfort while helping to provide protection from frontal exposure during applications such as mixing/handling of chemicals in chemical industry and for EV battery manufacturing in particular, as well as paint spraying and tasks involving fine hazardous particles or composite materials.
- Thanks to the taped underarm seams on the upper body, it offers increased protection also for the chemicals/products/process against potential contamination from the wearer or from garment seams during the typical work activity.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Small |
| Gender | Unisex |
| Material | HDPE |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Attached Socks |
| Cuff Style | Attached Gloves |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction