missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrphostin B42, 99+%, Thermo Scientific™
Click to view available options
Quantity:
25 mg
100 mg
Tamaño de la unidad:
100mg
25mg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Tyrphostin B42 is used in acute lymphoblastic leukemia studies the compound has been reported to inhibit JAK2 from B cell precursors.
Solubility
Soluble in DMSO or ethanol
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Tyrphostin B42 is used in acute lymphoblastic leukemia studies the compound has been reported to inhibit JAK2 from B cell precursors.
Solubility
Soluble in DMSO or ethanol
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Especificaciones
Especificaciones
| Melting Point | 216°C to 220°C |
| Color | Green-Yellow |
| Quantity | 25 mg |
| Solubility Information | Soluble in DMSO or ethanol |
| Formula Weight | 294.31 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
| Chemical Name or Material | Tyrphostin B42 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido