missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ [Tyr0] Atrial Natriuretic Peptide II, Rat
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Species | Porcine |
| Purification Method | HPLC |
| Common Name | [Tyr126] ANP (126-149), rat |
| Content And Storage | -30°C to -10°C |
| Format | Lyophilized Powder |
| Molecular Weight (g/mol) | 2549.79 |
| Product Type | [Tyr0] Atrial Natriuretic Peptide II, rat |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | [Tyr0] Atriopeptin II |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction